siRNA Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032558.2 | NP_115947.2 | 3, 4 | 535 | ||
XM_006717305.3 | XP_006717368.1 | 3, 4 | 510 | |||
XM_011519114.2 | XP_011517416.1 | 3, 4 | 226 | |||
XM_017015217.1 | XP_016870706.1 | 3, 4 | 336 | |||
XM_017015218.1 | XP_016870707.1 | 3, 4 | 263 | |||
XM_017015219.1 | XP_016870708.1 | 3, 4 | 292 | |||
XM_017015220.1 | XP_016870709.1 | 3, 4 | 202 | |||
XM_017015221.1 | XP_016870710.1 | 3, 4 | 226 | |||
GenBank mRNA | AK300615.1 | - | 2, 3 | 107 | ||
BC106062.1 | - | 3, 4 | 272 | 6162855 | ||
BU507687.1 | - | 3, 4 | 512 | |||
BX359483.2 | - | 3, 4 | 542 |
Gene Symbol: MFSD14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002894.1 | 3, 4 | 480 | |||
GenBank mRNA | BC005058.1 | - | 3, 4 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
