siRNA Details
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099285.1 | NP_001092755.1 | 4 | 427 | ||
NM_002823.4 | NP_002814.3 | 4 | 424 | |||
XM_011511518.1 | XP_011509820.1 | 4 | 1157 | |||
XM_011511519.1 | XP_011509821.1 | 4 | 1154 | |||
XM_011511520.1 | XP_011509822.1 | 4 | 1056 | |||
XM_011511521.1 | XP_011509823.1 | 4 | 1053 | |||
XM_011511522.2 | XP_011509824.1 | 3 | 1917 | |||
GenBank mRNA | AF348514.1 | - | 4 | 242 | ||
AF452640.1 | - | 4 | 724 | |||
AK223605.1 | - | 4 | 419 | |||
AK314106.1 | - | 4 | 419 | |||
AY169282.1 | - | 4 | 242 | |||
BC003510.1 | - | 4 | 407 | 3610808 | ||
BC022433.1 | - | 4 | 423 | 4277490 | ||
BC034921.1 | - | 4 | 415 | 4335961 | ||
BC051265.1 | - | 4 | 407 | 3832454 | ||
BC066905.1 | - | 4 | 345 | 4830132 | ||
BC070480.1 | - | 4 | 409 | 6205582 | ||
BC071647.1 | - | 4 | 402 | 5112610 | ||
BC071879.1 | - | 4 | 404 | 6425103 | ||
BC098420.1 | - | 4 | 409 | 5017164 | ||
BM839980.1 | - | 4 | 424 | |||
BX647606.1 | - | 1 | 1551 | |||
M14483.1 | - | 4 | 374 | |||
M14630.1 | - | 4 | 422 | |||
M14794.1 | - | 4 | 310 | |||
M26708.1 | - | 4 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
