siRNA Details
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099285.1 | NP_001092755.1 | 3 | 355 | ||
NM_002823.4 | NP_002814.3 | 3 | 352 | |||
XM_011511518.1 | XP_011509820.1 | 3 | 1085 | |||
XM_011511519.1 | XP_011509821.1 | 3 | 1082 | |||
XM_011511520.1 | XP_011509822.1 | 3 | 984 | |||
XM_011511521.1 | XP_011509823.1 | 3 | 981 | |||
GenBank mRNA | AF348514.1 | - | 3 | 170 | ||
AF452640.1 | - | 3 | 652 | |||
AK223605.1 | - | 3 | 347 | |||
AK314106.1 | - | 3 | 347 | |||
AY169282.1 | - | 3 | 170 | |||
BC003510.1 | - | 3 | 335 | 3610808 | ||
BC022433.1 | - | 3 | 351 | 4277490 | ||
BC034921.1 | - | 3 | 343 | 4335961 | ||
BC051265.1 | - | 3 | 335 | 3832454 | ||
BC066905.1 | - | 3 | 273 | 4830132 | ||
BC070480.1 | - | 3 | 337 | 6205582 | ||
BC071647.1 | - | 3 | 330 | 5112610 | ||
BC071879.1 | - | 3 | 332 | 6425103 | ||
BC098420.1 | - | 3 | 337 | 5017164 | ||
BM839980.1 | - | 3 | 352 | |||
BX647606.1 | - | 1 | 1020 | |||
M14483.1 | - | 3 | 302 | |||
M14630.1 | - | 3 | 350 | |||
M14794.1 | - | 3 | 238 | |||
M26708.1 | - | 3 | 325 |
Gene Symbol: LOC100506248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011520895.2 | XP_011519197.1 | 1 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
