siRNA Details
Gene Symbol: PTMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099285.1 | NP_001092755.1 | 3 | 379 | ||
NM_002823.4 | NP_002814.3 | 3 | 376 | |||
XM_011511518.1 | XP_011509820.1 | 3 | 1109 | |||
XM_011511519.1 | XP_011509821.1 | 3 | 1106 | |||
XM_011511520.1 | XP_011509822.1 | 3 | 1008 | |||
XM_011511521.1 | XP_011509823.1 | 3 | 1005 | |||
GenBank mRNA | AF348514.1 | - | 3 | 194 | ||
AF452640.1 | - | 3 | 676 | |||
AK223605.1 | - | 3 | 371 | |||
AK314106.1 | - | 3 | 371 | |||
AY169282.1 | - | 3 | 194 | |||
BC003510.1 | - | 3 | 359 | 3610808 | ||
BC022433.1 | - | 3 | 375 | 4277490 | ||
BC034921.1 | - | 3 | 367 | 4335961 | ||
BC051265.1 | - | 3 | 359 | 3832454 | ||
BC066905.1 | - | 3 | 297 | 4830132 | ||
BC070480.1 | - | 3 | 361 | 6205582 | ||
BC071647.1 | - | 3 | 354 | 5112610 | ||
BC071879.1 | - | 3 | 356 | 6425103 | ||
BC098420.1 | - | 3 | 361 | 5017164 | ||
BM839980.1 | - | 3 | 376 | |||
BX647606.1 | - | 1 | 1044 | |||
M14483.1 | - | 3 | 326 | |||
M14630.1 | - | 3 | 374 | |||
M14794.1 | - | 3 | 262 | |||
M26708.1 | - | 3 | 349 |
Gene Symbol: CLSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190481.1 | NP_001177410.1 | 9 | 1894 | ||
NM_022111.3 | NP_071394.2 | 10 | 2086 | |||
XM_011541938.2 | XP_011540240.1 | 10 | 2109 | |||
XM_011541939.2 | XP_011540241.1 | 10 | 2103 | |||
XM_011541940.2 | XP_011540242.1 | 10 | 2103 | |||
XM_011541941.2 | XP_011540243.1 | 9 | 1941 | |||
GenBank mRNA | AF297866.1 | - | 10 | 2086 | ||
BC038991.2 | - | 10 | 2112 | |||
BC113116.1 | - | 10 | 2079 | 7961377 | ||
BC115025.1 | - | 9 | 1887 | 7961363 | ||
BC115026.1 | - | 10 | 2079 | 7961366 | ||
BC137279.1 | - | 10 | 2065 | 9021276 | ||
BC140789.1 | - | 10 | 2065 | 9021657 |
Gene Symbol: LOC102467080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104658.1 | 2 | 426 | |||
NR_104659.1 | 2 | 184 | ||||
GenBank mRNA | AB371498.1 | - | 2 | 184 | ||
BM724720.1 | - | 2 | 426 | |||
CD357266.1 | - | 2 | 81 |
Gene Symbol: RGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004761.4 | NP_004752.1 | 2 | 703 | ||
GenBank mRNA | AB012295.1 | - | 1 | 55 | ||
AL050259.1 | - | 2 | 276 | |||
BC032681.1 | - | 2 | 258 | 5585507 | ||
BC040989.1 | - | 2 | 293 |
Gene Symbol: STRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516137.2 | XP_011514439.1 | 5 | 1179 | ||
XM_011516138.2 | XP_011514440.1 | 5 | 564 | |||
XM_011516139.2 | XP_011514441.1 | 6 | 808 | |||
XM_017012150.1 | XP_016867639.1 | 5 | 1179 |
Gene Symbol: ZCCHC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185059.1 | NP_001171988.1 | 13 | 2705 | ||
NM_001185074.1 | NP_001172003.1 | 9 | 2446 | |||
NM_024617.3 | NP_078893.2 | 13 | 2815 | |||
XM_005252207.3 | XP_005252264.1 | 13 | 2767 | |||
XM_005252208.2 | XP_005252265.1 | 13 | 2767 | |||
XM_006717283.1 | XP_006717346.1 | 13 | 2767 | |||
XM_011519011.2 | XP_011517313.1 | 13 | 2767 | |||
XM_011519012.2 | XP_011517314.1 | 13 | 2715 | |||
XM_011519013.2 | XP_011517315.1 | 13 | 3384 | |||
XM_011519014.2 | XP_011517316.1 | 13 | 2986 | |||
XM_011519015.1 | XP_011517317.1 | 13 | 2767 | |||
XM_011519016.1 | XP_011517318.1 | 13 | 2767 | |||
XM_017015130.1 | XP_016870619.1 | 13 | 3383 | |||
XM_017015131.1 | XP_016870620.1 | 13 | 3025 | |||
XM_017015132.1 | XP_016870621.1 | 14 | 2930 | |||
GenBank mRNA | AB051498.1 | - | 5 | 1353 | ||
AK055948.1 | - | 1 | 665 | |||
AL832026.1 | - | 9 | 2413 | |||
AL832193.1 | - | 14 | 2860 | |||
BC144523.1 | - | 12 | 2606 | |||
BX641077.1 | - | 12 | 4165 | |||
CR933643.1 | - | 13 | 2682 | |||
CR933644.1 | - | 14 | 2863 | |||
CR936608.1 | - | 14 | 2860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
