siRNA Details
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000566.3 | NP_000557.1 | 4 | 457 | ||
XM_005244957.3 | XP_005245014.1 | 4 | 460 | |||
XM_005244958.4 | XP_005245015.1 | 2 | 198 | |||
GenBank mRNA | AK291451.1 | - | 4 | 440 | ||
AK291502.1 | - | 4 | 457 | |||
BC032634.1 | - | 4 | 419 | 5248549 | ||
BC152383.1 | - | 4 | 410 | 100013442 | ||
DC320241.1 | - | 4 | 457 | |||
DQ786309.1 | - | 2 | 134 | |||
L03418.1 | - | 4 | 407 | |||
X14355.1 | - | 4 | 462 | |||
X14356.1 | - | 4 | 443 |
Gene Symbol: FCGR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004340.3 | NP_001004340.2 | 2 | 181 | ||
NM_001017986.3 | NP_001017986.1 | 4 | 457 | |||
NM_001244910.1 | NP_001231839.1 | 4 | 457 | |||
NR_045213.1 | 4 | 457 | ||||
XM_017000661.1 | XP_016856150.1 | 4 | 460 | |||
XM_017000662.1 | XP_016856151.1 | 3 | 202 | |||
GenBank mRNA | AK307244.1 | - | 4 | 457 | ||
AK308886.1 | - | 4 | 443 | |||
L03419.1 | - | 4 | 407 | |||
L03420.1 | - | 2 | 131 |
Gene Symbol: FCGR1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027484.2 | 4 | 457 | |||
GenBank mRNA | EB386870.1 | - | 2 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
