siRNA Details
Gene Symbol: FCGR1CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027484.2 | 6 | 1018 | |||
GenBank mRNA | EB386870.1 | - | 4 | 670 |
Gene Symbol: FCGR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291502.1 | - | 6 | 1016 |
Gene Symbol: FCGR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004340.3 | NP_001004340.2 | 3 | 458 | ||
NM_001017986.3 | NP_001017986.1 | 5 | 734 | |||
NR_045213.1 | 6 | 1019 | ||||
XM_017000661.1 | XP_016856150.1 | 5 | 737 | |||
XM_017000662.1 | XP_016856151.1 | 4 | 479 | |||
GenBank mRNA | L03419.1 | - | 5 | 684 | ||
L03420.1 | - | 3 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
