siRNA Details
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 2 | 176 | ||
GenBank mRNA | AF098306.1 | - | 2 | 152 | ||
AJ238277.1 | - | 2 | 155 | |||
AY920732.1 | - | 2 | 116 | |||
AY920733.1 | - | 2 | 116 | |||
AY920735.1 | - | 2 | 116 | |||
AY920736.1 | - | 2 | 116 | |||
AY920737.1 | - | 2 | 116 | |||
AY920738.1 | - | 2 | 116 | |||
AY920739.1 | - | 2 | 116 | |||
AY920741.1 | - | 2 | 116 | |||
AY920742.1 | - | 2 | 116 | |||
AY920743.1 | - | 2 | 116 | |||
AY920744.1 | - | 2 | 116 | |||
AY920745.1 | - | 2 | 116 | |||
AY920746.1 | - | 2 | 116 | |||
BC054023.1 | - | 2 | 177 | 6648369 | ||
BC069393.1 | - | 2 | 152 | 7262682 | ||
BC069816.1 | - | 2 | 152 | 7262670 | ||
CB115288.1 | - | 2 | 176 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 2 | 176 | ||
GenBank mRNA | CB110020.1 | - | 2 | 176 | ||
CB113891.1 | - | 2 | 176 |
Gene Symbol: SPANXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032461.3 | NP_115850.2 | 2 | 237 | ||
GenBank mRNA | AF098307.1 | - | 2 | 237 | ||
AJ457796.1 | - | 2 | 237 | |||
BC034472.1 | - | 2 | 235 | 4824918 | ||
BC137533.1 | - | 2 | 227 | 9021533 | ||
BC137536.1 | - | 2 | 227 | 9021536 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA412605.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
