siRNA Details
Gene Symbol: SPANXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032461.3 | NP_115850.2 | 2 | 308 | ||
GenBank mRNA | AF098307.1 | - | 2 | 308 | ||
AJ457796.1 | - | 2 | 308 | |||
BC034472.1 | - | 2 | 306 | 4824918 | ||
BC137533.1 | - | 2 | 298 | 9021533 | ||
BC137536.1 | - | 2 | 298 | 9021536 |
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ238277.1 | - | 2 | 226 | ||
AK225988.1 | - | 2 | 246 | |||
AY920737.1 | - | 2 | 187 | |||
BC054023.1 | - | 2 | 248 | 6648369 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 2 | 280 | ||
GenBank mRNA | AA412605.1 | - | 2 | 243 |
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ457795.1 | - | 2 | 223 | ||
BC005382.1 | - | 2 | 280 | 3935644 | ||
BC062672.1 | - | 2 | 461 | 4047937 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
