siRNA Details
Gene Symbol: LLPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032338.3 | NP_115714.1 | 2, 3 | 268 | ||
GenBank mRNA | AK057947.1 | - | 1, 2 | 181 | ||
AK091568.1 | - | 2, 3 | 268 | |||
BC006002.1 | - | 2, 3 | 264 | 4247279 | ||
BC070232.1 | - | 2, 3 | 260 | 4594274 | ||
BC107780.1 | - | 2, 3 | 269 | 6698895 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
