siRNA Details
Gene Symbol: CYSRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199001.3 | NP_945352.3 | 2 | 921 | ||
GenBank mRNA | BE563091.1 | - | 1 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
