siRNA Details
Gene Symbol: ZNF471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321768.1 | NP_001308697.1 | 4 | 3541 | ||
NM_020813.3 | NP_065864.2 | 5 | 3668 | |||
XM_011527148.1 | XP_011525450.1 | 5 | 6495 | |||
XM_017027032.1 | XP_016882521.1 | 4 | 6368 | |||
GenBank mRNA | AB037817.1 | - | 2 | 3310 | ||
AL831845.1 | - | 5 | 3668 | |||
BC016830.1 | - | 1 | 142 | |||
BC028227.1 | - | 1 | 284 | |||
BC125221.1 | - | 5 | 3626 | 40124078 | ||
BC125222.1 | - | 5 | 3626 | 40124080 |
Gene Symbol: LOC100131257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034022.1 | 1 | 8621 |
Gene Symbol: LOC55338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130914.1 | 3 | 1854 | |||
GenBank mRNA | AK002104.1 | - | 2 | 2548 | ||
AK002154.1 | - | 2 | 1775 | |||
AK023417.1 | - | 3 | 1855 | |||
AK024354.1 | - | 1 | 1501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
