siRNA Details
Gene Symbol: SSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278691.1 | NP_001265620.1 | 6 | 515 | ||
NM_005635.3 | NP_005626.1 | 5 | 429 | |||
GenBank mRNA | BC001003.2 | - | 5 | 374 | 3445470 | |
BC125151.1 | - | 5 | 372 | 40120554 | ||
BC128611.1 | - | 5 | 371 | 6159943 | ||
BC133693.1 | - | 5 | 372 | 40120550 | ||
BC150487.2 | - | 5 | 376 | 40120555 | ||
BI831262.1 | - | 5 | 427 | |||
X86174.1 | - | 5 | 384 |
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021015.3 | NP_066295.3 | 6 | 469 | ||
NM_175723.1 | NP_783729.1 | 5 | 346 | |||
XM_011543949.2 | XP_011542251.1 | 5 | 381 | |||
XM_017029758.1 | XP_016885247.1 | 5 | 381 | |||
GenBank mRNA | BC016640.2 | - | 5 | 435 | 3923927 | |
BE896093.1 | - | 5 | 449 | |||
U90842.1 | - | 4 | 293 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
