siRNA Details
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022114.1 | - | 5 | 1122 | ||
AK311390.1 | - | 5 | 1025 | |||
BC041839.1 | - | 4 | 1674 | |||
BX648537.1 | - | 5 | 1169 | |||
BX648730.1 | - | 6 | 5994 |
Gene Symbol: ACOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021592.1 | XP_016877081.1 | 1 | 17 |
Gene Symbol: CCDC180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627453.1 | - | 1 | 332 |
Gene Symbol: COG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832190.1 | - | 8 | 2331 |
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648946.1 | - | 1 | 3075 |
Gene Symbol: STRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014894.1 | XP_016870383.1 | 1 | 315 | ||
XM_017014895.1 | XP_016870384.1 | 1 | 315 |
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019069.3 | NP_061942.2 | 1 | 2574 | ||
GenBank mRNA | BC043494.1 | - | 1 | 2163 | 6056052 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
