siRNA Details
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 5 | 443 | |||
GenBank mRNA | AK289851.1 | - | 4 | 369 | ||
CN312642.1 | - | 2 | 233 | |||
X83301.1 | - | 8 | 891 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 4 | 471 | |||
GenBank mRNA | BC034017.1 | - | 4 | 472 | ||
BC067351.1 | - | 2 | 169 | |||
BC140728.1 | - | 3 | 294 | |||
BC171739.1 | - | 3 | 294 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027386.2 | 4 | 449 | |||
GenBank mRNA | BT006760.1 | - | 2 | 124 | ||
DT218991.1 | - | 4 | 369 | |||
X83299.1 | - | 4 | 400 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 2 | 118 | |||
NR_133000.1 | 2 | 118 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 2 | 181 | |||
GenBank mRNA | AW500063.1 | - | 2 | 114 | ||
BC104473.1 | - | 2 | 181 | |||
CF141910.1 | - | 2 | 187 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 4 | 358 | |||
NR_132997.1 | 6 | 617 | ||||
NR_132998.1 | 3 | 285 | ||||
GenBank mRNA | BC144584.1 | - | 4 | 294 | ||
BC144585.1 | - | 3 | 221 | |||
BM553931.1 | - | 6 | 617 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 1 | 75 | |||
NR_029426.1 | 4 | 367 | ||||
GenBank mRNA | AK094021.1 | - | 5 | 773 | ||
AK097091.1 | - | 5 | 747 | |||
AK289832.1 | - | 4 | 449 | |||
AK315849.1 | - | 4 | 440 | |||
X75940.1 | - | 1 | 76 | |||
X83300.1 | - | 2 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
