siRNA Details
Gene Symbol: KRTAP5-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012708.2 | NP_001012726.1 | 1 | 504 | ||
GenBank mRNA | AB126072.1 | - | 1 | 504 | ||
AJ628246.1 | - | 1 | 214 | |||
AY597812.1 | - | 1 | 426 |
Gene Symbol: KRTAP5-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012709.1 | NP_001012727.1 | 681 | |||
GenBank mRNA | AB126073.1 | - | 681 |
Gene Symbol: KRTAP5-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001480.2 | NP_001001480.2 | 1 | 548 | ||
GenBank mRNA | AB126074.1 | - | 2 | 558 | ||
AJ628247.1 | - | 2 | 530 |
Gene Symbol: KRTAP5-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012416.1 | NP_001012416.1 | 1 | 354 | ||
GenBank mRNA | AB126075.1 | - | 1 | 354 | ||
BC130399.1 | - | 1 | 326 | 40146430 | ||
BC133035.1 | - | 1 | 326 | 40147057 |
Gene Symbol: KRTAP5-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012503.1 | NP_001012521.1 | 1 | 358 | ||
GenBank mRNA | AB126076.1 | - | 1 | 358 | ||
AJ628243.1 | - | 1 | 31 | |||
BC136967.1 | - | 1 | 347 | 9020959 | ||
BC136968.1 | - | 1 | 347 | 9020960 |
Gene Symbol: KRTAP5-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021046.2 | NP_066384.2 | 1 | 334 | ||
GenBank mRNA | AB126077.1 | - | 1 | 334 | ||
AY360461.1 | - | 1 | 303 | |||
BC119736.1 | - | 3 | 387 |
Gene Symbol: KRTAP5-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005553.3 | NP_005544.4 | 1 | 517 | ||
GenBank mRNA | AB126078.1 | - | 1 | 313 | ||
BC069531.1 | - | 1 | 279 | 7262098 | ||
BC101744.1 | - | 1 | 421 | 8069250 | ||
X63755.1 | - | 1 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
