siRNA Details
Gene Symbol: KRTAP5-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012709.1 | NP_001012727.1 | 78 | |||
GenBank mRNA | AB126073.1 | - | 78 |
Gene Symbol: KRTAP5-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012710.1 | NP_001012728.1 | 1 | 58 | ||
GenBank mRNA | AB126079.1 | - | 1 | 58 | ||
BC137550.1 | - | 1 | 58 | 9021550 | ||
BC137566.1 | - | 1 | 58 | 9021566 |
Gene Symbol: KRTAP5-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004325.1 | NP_001004325.1 | 1 | 77 | ||
GenBank mRNA | AB126071.1 | - | 1 | 85 | ||
AJ628245.1 | - | 1 | 77 | |||
BC132882.1 | - | 1 | 56 | 40146904 | ||
BC132908.1 | - | 1 | 56 | 40146930 |
Gene Symbol: KRTAP5-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001480.2 | NP_001001480.2 | 1 | 71 | ||
GenBank mRNA | AB126074.1 | - | 1 | 99 | ||
AJ628247.1 | - | 1 | 71 |
Gene Symbol: KRTAP5-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012416.1 | NP_001012416.1 | 1 | 84 | ||
GenBank mRNA | AB126075.1 | - | 1 | 84 | ||
BC130399.1 | - | 1 | 56 | 40146430 | ||
BC133035.1 | - | 1 | 56 | 40147057 |
Gene Symbol: KRTAP5-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021046.2 | NP_066384.2 | 1 | 64 | ||
GenBank mRNA | AB126077.1 | - | 1 | 64 | ||
AY360461.1 | - | 1 | 33 | |||
BC119736.1 | - | 1 | 33 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
