siRNA Details
Gene Symbol: KRTAP5-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005405.2 | NP_001005405.1 | 1 | 71 | ||
GenBank mRNA | AB126080.1 | - | 1 | 71 | ||
BC130498.1 | - | 1 | 59 | |||
BC130502.1 | - | 1 | 59 |
Gene Symbol: KRTAP5-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001480.2 | NP_001001480.2 | 1 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
