siRNA Details
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204118.1 | NP_001191047.1 | 14 | 1898 | ||
NM_207390.3 | NP_997273.3 | 13 | 1788 | |||
NR_109784.1 | 14 | 1961 | ||||
NR_109785.1 | 13 | 1746 | ||||
XM_017026785.1 | XP_016882274.1 | 15 | 2442 | |||
XM_017026786.1 | XP_016882275.1 | 15 | 2306 | |||
XM_017026787.1 | XP_016882276.1 | 16 | 2661 | |||
XM_017026788.1 | XP_016882277.1 | 15 | 2418 | |||
XM_017026789.1 | XP_016882278.1 | 11 | 2120 | |||
XM_017026790.1 | XP_016882279.1 | 14 | 2394 | |||
XM_017026791.1 | XP_016882280.1 | 13 | 2243 |
Gene Symbol: FAM57B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097527.1 | - | 2, 3 | 705 |
Gene Symbol: GPR132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA714520.1 | - | 1 | 101 |
Gene Symbol: IDNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717111.3 | XP_006717174.1 | 1 | 176 |
Gene Symbol: PIK3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090567.1 | - | 1 | 1325 |
Gene Symbol: TPTE2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002815.2 | 3 | 1633 | |||
GenBank mRNA | AL122103.1 | - | 1 | 495 | ||
AL137411.1 | - | 7 | 591 | |||
BC045761.1 | - | 3 | 1632 | |||
DN831683.1 | - | 3 | 188 |
Gene Symbol: UGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712088.1 | XP_006712151.1 | 3 | 443 | ||
XM_017004857.1 | XP_016860346.1 | 3 | 443 |
Gene Symbol: ZNF587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026676.1 | - | 2 | 1239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
