siRNA Details
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 3 | 311 | |||
GenBank mRNA | AW500063.1 | - | 3 | 244 | ||
BC104473.1 | - | 3 | 311 | |||
CF141910.1 | - | 3 | 317 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 5 | 460 | |||
GenBank mRNA | AK290693.1 | - | 5 | 427 | ||
BC147000.1 | - | 5 | 414 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 5 | 601 | |||
GenBank mRNA | BC034017.1 | - | 5 | 602 | ||
BC067351.1 | - | 3 | 299 | |||
BC140728.1 | - | 4 | 424 | |||
BC171739.1 | - | 4 | 424 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027386.2 | 5 | 579 | |||
GenBank mRNA | AK289965.1 | - | 4 | 551 | ||
BT006760.1 | - | 3 | 254 | |||
DT218991.1 | - | 5 | 499 | |||
U21555.1 | - | LIT2100 5G4 pseudogene |
2 | 42 | ||
X83299.1 | - | 5 | 530 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 3 | 248 | |||
NR_133000.1 | 3 | 248 | ||||
GenBank mRNA | AW955360.1 | - | 2 | 76 | ||
DQ583004.1 | - | 1 | 12 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 5 | 488 | |||
NR_132997.1 | 7 | 747 | ||||
NR_132998.1 | 4 | 415 | ||||
GenBank mRNA | BC144584.1 | - | 5 | 424 | ||
BC144585.1 | - | 4 | 351 | |||
BM553931.1 | - | 7 | 747 | |||
DA269273.1 | - | 4 | 245 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 2 | 205 | |||
NR_029426.1 | 5 | 497 | ||||
GenBank mRNA | AK094021.1 | - | 6 | 902 | ||
AK097091.1 | - | 6 | 877 | |||
AK289832.1 | - | 5 | 579 | |||
AK315849.1 | - | 5 | 570 | |||
BC131740.1 | - | 3 | 311 | |||
X75940.1 | - | 2 | 206 | |||
X83300.1 | - | 3 | 320 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 6 | 573 | |||
GenBank mRNA | AK289851.1 | - | 5 | 499 | ||
CN312642.1 | - | 3 | 363 | |||
X83301.1 | - | 6 | 573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
