siRNA Details
Gene Symbol: FAM177B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324080.1 | NP_001311009.1 | 2 | 225 | ||
NM_207468.2 | NP_997351.2 | 3 | 327 | |||
NR_136691.1 | 3 | 327 | ||||
XM_017001279.1 | XP_016856768.1 | 2 | 769 | |||
XM_017001280.1 | XP_016856769.1 | 3 | 283 | |||
XM_017001281.1 | XP_016856770.1 | 3 | 852 | |||
XM_017001282.1 | XP_016856771.1 | 3 | 308 | |||
XM_017001283.1 | XP_016856772.1 | 2 | 101 | |||
XM_017001284.1 | XP_016856773.1 | 2 | 101 | |||
XM_017001285.1 | XP_016856774.1 | 2 | 101 | |||
GenBank mRNA | AK125494.1 | - | 3 | 327 | ||
CA848039.1 | - | 2 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
