siRNA Details
Gene Symbol: ANKRD20A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012419.2 | NP_001012419.1 | 13 | 1327 | ||
XM_005272525.3 | XP_005272582.1 | 13 | 1580 | |||
XM_011543860.2 | XP_011542162.1 | 13 | 1584 | |||
XM_011543861.2 | XP_011542163.1 | 13 | 1588 | |||
GenBank mRNA | BC140809.1 | - | 13 | 1324 | 9021677 | |
BC171793.1 | - | 13 | 1324 | 9054447 |
Gene Symbol: ANKRD20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032250.3 | NP_115626.2 | 13 | 1361 | ||
XM_005272532.4 | XP_005272589.1 | 13 | 1295 | |||
XM_011545682.2 | XP_011543984.1 | 13 | 1298 | |||
GenBank mRNA | AL136793.1 | - | 13 | 1362 |
Gene Symbol: ANKRD20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012421.1 | NP_001012421.1 | 13 | 1361 | ||
XM_011517877.2 | XP_011516179.1 | 13 | 1361 | |||
XM_011517878.2 | XP_011516180.1 | 13 | 1361 | |||
XM_011517879.2 | XP_011516181.1 | 13 | 1311 |
Gene Symbol: ANKRD20A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098805.1 | NP_001092275.1 | 13 | 1361 |
Gene Symbol: ANKRD20A5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040113.1 | 13 | 1546 |
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 11 | 1066 |
Gene Symbol: ANKRD20A9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027995.1 | 1 | 30353 | |||
GenBank mRNA | BJ991373.1 | - | 6 | 556 |
Gene Symbol: LOC102723382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546266.2 | XP_011544568.1 | 5 | 442 |
Gene Symbol: LOC105379417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030104.1 | XP_016885593.1 | 8 | 1878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
