siRNA Details
Gene Symbol: ANKRD20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032250.3 | NP_115626.2 | 4 | 680 | ||
XM_005272532.4 | XP_005272589.1 | 4 | 617 | |||
XM_011545682.2 | XP_011543984.1 | 4 | 617 | |||
GenBank mRNA | AL136793.1 | - | 4 | 681 |
Gene Symbol: ANKRD20A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098805.1 | NP_001092275.1 | 4 | 680 | ||
GenBank mRNA | AK307323.1 | - | 5 | 1058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
