siRNA Details
Gene Symbol: CD99L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184808.1 | NP_001171737.1 | 2 | 338 | ||
NM_001242614.1 | NP_001229543.1 | 2 | 338 | |||
NM_031462.3 | NP_113650.2 | 2 | 338 | |||
NM_134445.3 | NP_604394.1 | 2 | 338 | |||
NM_134446.3 | NP_604395.1 | 2 | 338 | |||
XM_011531202.2 | XP_011529504.1 | 2 | 239 | |||
XM_017029890.1 | XP_016885379.1 | 2 | 202 | |||
XM_017029891.1 | XP_016885380.1 | 2 | 336 | |||
GenBank mRNA | AK023776.1 | - | 1 | 24 | ||
AK125020.1 | - | 2 | 149 | |||
AK290200.1 | - | 2 | 114 | |||
AK291375.1 | - | 2 | 151 | |||
AK293244.1 | - | 2 | 144 | |||
AK295217.1 | - | 2 | 156 | |||
AL136580.1 | - | 2 | 228 | |||
AY078165.1 | - | 2 | 146 | |||
AY078166.1 | - | 2 | 146 | |||
AY078167.1 | - | 2 | 146 | |||
AY358837.1 | - | 2 | 211 | |||
BC025729.1 | - | 2 | 152 | 5203087 | ||
BC030536.1 | - | 2 | 198 | 5242536 | ||
DC323066.1 | - | 2 | 338 | |||
KU178797.1 | - | 2 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
