siRNA Details
Gene Symbol: GPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321525.1 | NP_001308454.1 | 10 | 1371 | ||
NM_001321526.1 | NP_001308455.1 | 10 | 1461 | |||
NM_001321527.1 | NP_001308456.1 | 10 | 1425 | |||
NM_001321528.1 | NP_001308457.1 | 10 | 1425 | |||
NM_001321531.1 | NP_001308460.1 | 11 | 1432 | |||
NM_207328.3 | NP_997211.2 | 10 | 1461 | |||
XM_005263888.3 | XP_005263945.1 | 10 | 1136 | |||
XM_006712308.3 | XP_006712371.1 | 10 | 1136 | |||
XM_006712311.3 | XP_006712374.1 | 10 | 1136 | |||
XM_006712312.3 | XP_006712375.1 | 10 | 1136 | |||
XM_006712313.3 | XP_006712376.1 | 10 | 1136 | |||
XM_017003424.1 | XP_016858913.1 | 10 | 1075 | |||
XM_017003425.1 | XP_016858914.1 | 10 | 1136 | |||
XM_017003426.1 | XP_016858915.1 | 10 | 1136 | |||
GenBank mRNA | AK122580.1 | - | 4 | 1271 | ||
AK131194.1 | - | 11 | 1432 | |||
AK131197.1 | - | 4 | 480 | |||
AK302887.1 | - | 9 | 1534 | |||
AK302918.1 | - | 9 | 1545 | |||
BC068596.1 | - | 10 | 1132 | 4837765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
