siRNA Details
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 32 | 6493 | ||
NM_001265611.1 | NP_001252540.1 | 30 | 5644 | |||
NM_014994.2 | NP_055809.2 | 31 | 6475 | |||
NR_049761.1 | 30 | 6285 | ||||
NR_049762.1 | 29 | 6236 | ||||
XM_006720438.1 | XP_006720501.1 | 30 | 6292 | |||
XM_006720439.2 | XP_006720502.1 | 18 | 4567 | |||
XM_011521383.1 | XP_011519685.1 | 31 | 6310 | |||
XM_017022017.1 | XP_016877506.1 | 30 | 6586 | |||
GenBank mRNA | AB011168.2 | - | 32 | 6403 | ||
AB177850.1 | - | 23 | 5317 | |||
AL833267.1 | - | 31 | 6395 | |||
BX648786.1 | - | 31 | 6426 |
Gene Symbol: ACAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036445.1 | - | 10 | 3382 |
Gene Symbol: ACAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222635.1 | - | 7 | 1579 |
Gene Symbol: C1RL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091792.1 | - | 8 | 1887 |
Gene Symbol: COA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023237.1 | - | 1 | 1663 |
Gene Symbol: FLJ36777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094096.1 | - | 1 | 2111 |
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005258682.4 | XP_005258739.3 | 7 | 1673 |
Gene Symbol: HELZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023724.1 | - | 4 | 1790 | ||
BC104974.1 | - | 3 | 1268 | |||
BC112023.1 | - | 3 | 1268 |
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246334.4 | XP_005246391.1 | 6 | 2328 | ||
XM_006712320.3 | XP_006712383.1 | 6 | 2410 |
Gene Symbol: LINC00954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022396.1 | - | 5 | 2028 | ||
AK097927.1 | - | 6 | 2121 |
Gene Symbol: LOC100130370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL162045.1 | - | 1 | 1249 |
Gene Symbol: LOC101929007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC033373.1 | - | 1 | 1682 |
Gene Symbol: NOTCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063815.1 | - | 11 | 2592 |
Gene Symbol: NPHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055893.1 | - | 1 | 1300 |
Gene Symbol: NUP188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005407.1 | - | 19 | 2723 |
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198887.2 | NP_942590.1 | 8 | 3385 | ||
XM_005266960.4 | XP_005267017.1 | 8 | 4615 | |||
XM_005266961.3 | XP_005267018.1 | 9 | 3435 | |||
XM_005266962.3 | XP_005267019.1 | 8 | 5655 | |||
XM_011535799.2 | XP_011534101.1 | 8 | 4697 | |||
GenBank mRNA | BC040121.1 | - | 6 | 3030 | ||
BC046637.1 | - | 1 | 3133 |
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 12 | 2328 | ||
XM_011544605.2 | XP_011542907.1 | 12 | 2213 | |||
XM_011544606.2 | XP_011542908.1 | 12 | 2264 | |||
XM_011544607.2 | XP_011542909.1 | 12 | 2226 | |||
XM_017013718.1 | XP_016869207.1 | 11 | 2079 | |||
XM_017013719.1 | XP_016869208.1 | 11 | 2049 | |||
XM_017013720.1 | XP_016869209.1 | 11 | 2164 | |||
GenBank mRNA | AK098787.1 | - | 8, 9 | 948 |
Gene Symbol: POLR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014399.1 | - | 8 | 1121 |
Gene Symbol: SLC12A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024494.1 | - | 1 | 1422 |
Gene Symbol: SLC7A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014331.3 | NP_055146.1 | 12 | 6375 | ||
GenBank mRNA | BC041925.1 | - | 1 | 2525 |
Gene Symbol: STX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095898.1 | - | 1 | 360 |
Gene Symbol: SULT2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096418.1 | - | 1 | 581 |
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK225445.1 | - | 10 | 2459 |
Gene Symbol: UBXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512670.2 | XP_011510972.1 | 11 | 5717 | ||
XM_011512671.2 | XP_011510973.1 | 10 | 5574 |
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032433.2 | NP_115809.1 | 12 | 2795 | ||
XM_005260098.3 | XP_005260155.1 | 13 | 3108 | |||
XM_006722924.2 | XP_006722987.1 | 12 | 2872 | |||
XM_011528362.2 | XP_011526664.1 | 12 | 2836 | |||
XM_011528363.2 | XP_011526665.1 | 14 | 3936 | |||
XM_011528365.2 | XP_011526667.1 | 14 | 3277 | |||
XM_011528366.2 | XP_011526668.1 | 13 | 3105 | |||
XM_011528367.2 | XP_011526669.1 | 13 | 3118 | |||
XM_011528368.2 | XP_011526670.1 | 13 | 3025 | |||
XM_011528369.2 | XP_011526671.1 | 13 | 3038 | |||
XM_011528371.2 | XP_011526673.1 | 6 | 3202 | |||
XM_011528372.2 | XP_011526674.1 | 5 | 2131 | |||
XM_017027367.1 | XP_016882856.1 | 12 | 2812 | |||
XM_017027368.1 | XP_016882857.1 | 11 | 2680 | |||
XM_017027369.1 | XP_016882858.1 | 13 | 4433 | |||
XM_017027370.1 | XP_016882859.1 | 13 | 4446 | |||
XM_017027371.1 | XP_016882860.1 | 13 | 3058 | |||
GenBank mRNA | AB058709.1 | - | 5 | 2109 | ||
AF372702.1 | - | 12 | 2752 | |||
AK095142.1 | - | 1 | 1677 | |||
AK122817.1 | - | 5 | 2676 | |||
AL832544.1 | - | 6 | 2286 | |||
BX647475.1 | - | 12 | 2756 |
Gene Symbol: ZNRF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046851.1 | 3 | 1972 | |||
GenBank mRNA | AK095488.1 | - | 3 | 1969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
