siRNA Details
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282729.1 | NP_001269658.1 | 6 | 3377 | ||
NM_001282730.1 | NP_001269659.1 | 6 | 3585 | |||
NM_001282731.1 | NP_001269660.1 | 6 | 3427 | |||
NM_001282732.1 | NP_001269661.1 | 5 | 2967 | |||
NM_014860.2 | NP_055675.1 | 6 | 3372 | |||
XM_005264672.4 | XP_005264729.1 | 6 | 3574 | |||
GenBank mRNA | AB018307.1 | - | 6 | 3114 |
Gene Symbol: AP4E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL355694.1 | - | 1 | 251 |
Gene Symbol: ARMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127341.1 | - | 1 | 2400 |
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128259.1 | - | 1 | 516 |
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098178.1 | - | 1 | 513 |
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006715785.3 | XP_006715848.1 | 1 | 240 | ||
XM_011515563.2 | XP_011513865.1 | 1 | 240 | |||
XM_017012672.1 | XP_016868161.1 | 1 | 240 |
Gene Symbol: CLSTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007022.1 | XP_016862511.1 | 17 | 6731 |
Gene Symbol: CNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093630.1 | - | 1 | 546 |
Gene Symbol: COTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125645.1 | - | 1 | 1146 |
Gene Symbol: CPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ181238.1 | - | 1 | 217 |
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054737.1 | - | 1 | 637 | ||
AK090414.1 | - | 11 | 4273 |
Gene Symbol: DBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016216.3 | NP_057300.2 | 8 | 2021 | ||
GenBank mRNA | AF180919.2 | - | 8 | 1975 |
Gene Symbol: DEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093613.1 | - | 4 | 487 |
Gene Symbol: DHX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054614.1 | - | 1 | 494 |
Gene Symbol: EFR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319099.1 | NP_001306028.1 | 23 | 4814 | ||
NM_014971.1 | NP_055786.1 | 23 | 4826 | |||
XM_011532701.2 | XP_011531003.1 | 23 | 4799 |
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR749455.1 | - | 2 | 3139 |
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126187.1 | - | 14 | 1980 |
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ438905.1 | - | 1 | 1854 |
Gene Symbol: GSTZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209360.1 | - | 1 | 1632 |
Gene Symbol: HEATR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256013.2 | XP_005256070.1 | 14 | 2736 | ||
XM_011523188.2 | XP_011521490.1 | 15 | 2811 | |||
XM_017023385.1 | XP_016878874.1 | 13 | 2874 | |||
XM_017023386.1 | XP_016878875.1 | 13 | 2679 |
Gene Symbol: HEXA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537989.1 | - | 2 | 872 |
Gene Symbol: HOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134670.1 | NP_001128142.1 | 3 | 1401 | ||
NM_138413.3 | NP_612422.2 | 7 | 1890 | |||
GenBank mRNA | BC011916.2 | - | 3 | 1401 | 4310241 | |
BC045550.1 | - | 7 | 1875 | 5274464 | ||
BC057821.1 | - | 3 | 1374 | 5263339 |
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031560.1 | - | 7 | 1088 |
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB037745.1 | - | 13 | 5008 |
Gene Symbol: LOC100128108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720812.3 | XP_006720875.1 | 1 | 668 |
Gene Symbol: LOC100130560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127462.1 | - | 1 | 2154 |
Gene Symbol: LOC101929140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120423.1 | 4 | 1557 | |||
GenBank mRNA | AK056182.1 | - | 4 | 1548 |
Gene Symbol: LOC105372129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130693.1 | - | 1 | 1868 |
Gene Symbol: LOC105376292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037833.1 | - | 4 | 730 |
Gene Symbol: LOC158402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056358.1 | - | 1 | 312 |
Gene Symbol: LRRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC064321.1 | - | 2 | 2043 |
Gene Symbol: LRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209379.1 | - | 7 | 3598 |
Gene Symbol: MORN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC093023.1 | - | 5 | 1348 | 30519946 |
Gene Symbol: NAA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719606.2 | XP_006719669.1 | 1 | 88 |
Gene Symbol: PGAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170543.1 | NP_001164014.1 | 6 | 2568 | ||
NM_001170544.1 | NP_001164015.1 | 6 | 2565 |
Gene Symbol: PHF20L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641040.1 | - | 1 | 3028 |
Gene Symbol: PTGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU616297.1 | - | 1 | 381 |
Gene Symbol: RBM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097195.1 | - | 2 | 459 |
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016120.3 | NP_057204.2 | 4 | 5262 | ||
NM_183353.2 | NP_899196.1 | 5 | 5324 | |||
GenBank mRNA | AK123088.1 | - | 1 | 1058 | ||
AK124877.1 | - | 1 | 420 | |||
BX648613.1 | - | 1 | 2580 |
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093848.1 | - | 2 | 485 |
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091238.1 | - | 1 | 896 |
Gene Symbol: SGPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003901.3 | NP_003892.2 | 15 | 2619 | ||
XM_005270263.1 | XP_005270320.1 | 15 | 2632 | |||
XM_006718053.1 | XP_006718116.1 | 16 | 2734 | |||
XM_011540316.2 | XP_011538618.1 | 15 | 2678 | |||
XM_011540317.1 | XP_011538619.1 | 15 | 2528 | |||
XM_011540318.1 | XP_011538620.1 | 14 | 2549 | |||
GenBank mRNA | AB033078.1 | - | 15 | 2595 | ||
AK023873.1 | - | 2 | 874 | |||
BC052991.1 | - | 15 | 2605 | 6150776 |
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC035480.1 | - | 2 | 1124 |
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 3 | 1917 | ||
GenBank mRNA | BC121148.1 | - | 2 | 1175 | 40121265 |
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 7 | 5022 | ||
NM_018299.4 | NP_060769.4 | 6 | 4989 | |||
NR_073119.1 | 6 | 5010 | ||||
NR_073120.1 | 6 | 4970 | ||||
NR_073121.1 | 5 | 4886 |
Gene Symbol: WBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128407.1 | - | 1 | 438 |
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010888.3 | NP_001010888.3 | 5 | 3192 | ||
XM_011530939.2 | XP_011529241.1 | 6 | 3437 | |||
XM_017029478.1 | XP_016884967.1 | 11 | 4703 | |||
XM_017029479.1 | XP_016884968.1 | 13 | 7996 | |||
XM_017029480.1 | XP_016884969.1 | 11 | 4737 | |||
XM_017029481.1 | XP_016884970.1 | 11 | 4732 | |||
XM_017029482.1 | XP_016884971.1 | 10 | 4642 | |||
XM_017029483.1 | XP_016884972.1 | 9 | 4438 | |||
XM_017029484.1 | XP_016884973.1 | 9 | 4070 | |||
GenBank mRNA | BC140767.1 | - | 5 | 3192 | 9021635 | |
BC171781.1 | - | 5 | 3192 | 9054435 | ||
BX647241.1 | - | 7 | 2942 |
Gene Symbol: ZNF250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109689.3 | NP_001103159.1 | 6 | 2625 | ||
NM_021061.4 | NP_066405.1 | 6 | 2640 | |||
XM_006716612.3 | XP_006716675.1 | 6 | 3140 | |||
XM_006716613.3 | XP_006716676.1 | 6 | 2679 |
Gene Symbol: ZNF555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832140.1 | - | 4 | 4445 |
Gene Symbol: ZNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209619.1 | - | 1 | 2149 |
Gene Symbol: ZNF70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126511.1 | - | 1 | 2942 |
Gene Symbol: ZNF785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040642.2 | - | 3 | 2878 | 4811661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
