siRNA Details
Gene Symbol: HLA-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005514.7 | NP_005505.2 | 3 | 623 | ||
XM_011514557.1 | XP_011512859.1 | 3 | 546 | |||
GenBank mRNA | AF189017.1 | - | 3 | 536 | ||
AF436098.1 | - | 3 | 536 | |||
AJ002675.1 | - | 2 | 463 | |||
AJ003063.1 | - | 2 | 463 | |||
AJ223602.1 | - | 2 | 463 | |||
AK300685.1 | - | 4 | 466 | |||
AK301008.1 | - | 3 | 443 | |||
AK310586.1 | - | 4 | 476 | |||
AK313911.1 | - | 3 | 557 | |||
HG514457.1 | - | 3 | 536 | |||
HM989017.1 | - | 2 | 463 | |||
KU204955.1 | - | 1 | 193 | |||
L33922.1 | - | 3 | 536 | |||
L47338.1 | - | 2 | 463 | |||
M16102.1 | - | 3 | 543 | |||
M27540.1 | - | 2 | 350 | |||
M32317.1 | - | 3 | 536 | |||
U18661.1 | - | 2 | 320 | |||
U21052.1 | - | 3 | 536 | |||
U21053.1 | - | 3 | 536 | |||
U29057.1 | - | 3 | 536 | |||
X64454.1 | - | 3 | 536 | |||
X91749.1 | - | 3 | 536 | |||
Y13567.1 | - | 3 | 536 |
Gene Symbol: HLA-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF168611.1 | - | 3 | 536 | ||
AF196489.1 | - | 4 | 536 | |||
AF405691.1 | - | 4 | 536 | |||
AJ010748.1 | - | 3 | 536 | |||
AM237285.2 | - | 3 | 536 | |||
D31817.1 | - | 3 | 536 | |||
D49820.1 | - | 3 | 463 | |||
D50852.1 | - | 3 | 536 | |||
D64145.1 | - | 3 | 536 | |||
D64148.1 | - | 3 | 536 | |||
D83030.1 | - | 4 | 536 | |||
FN662479.1 | - | 2 | 463 | |||
HG974569.1 | - | 4 | 536 | |||
M84171.1 | - | 3 | 536 | |||
M84173.1 | - | 3 | 536 | |||
M84386.1 | - | 4 | 536 | |||
U06487.1 | - | 3 | 536 | |||
U07230.1 | - | 2 | 440 | |||
U41386.1 | - | 3 | 536 | |||
U57028.1 | - | 3 | 536 | |||
X58536.1 | - | 4 | 551 | |||
X96582.1 | - | 3 | 536 | |||
Y09156.1 | - | 3 | 536 | |||
Z46809.1 | - | 3 | 536 | |||
Z47377.1 | - | 2 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
