siRNA Details
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033952.2 | NP_001029124.1 | 2 | 121 | ||
NM_001033953.2 | NP_001029125.1 | 2 | 121 | |||
NM_001741.2 | NP_001732.1 | 2 | 97 | |||
NR_125898.1 | 2 | 97 | ||||
XM_017018283.1 | XP_016873772.1 | 2 | 172 | |||
XM_017018284.1 | XP_016873773.1 | 2 | 196 | |||
GenBank mRNA | BC069684.1 | - | 2 | 97 | 7262564 | |
BC069704.1 | - | 2 | 97 | 7262481 | ||
BC069760.1 | - | 2 | 97 | 7262493 | ||
BC069778.1 | - | 2 | 97 | 7262505 | ||
BC093753.1 | - | 2 | 108 | 7939598 | ||
BC101599.1 | - | 2 | 84 | 8069105 | ||
BC143562.1 | - | 2 | 108 | 9052073 | ||
CD612754.1 | - | 1 | 30 | |||
M26095.1 | - | 2 | 80 | |||
M64486.1 | - | 2 | 85 | |||
X00356.1 | - | 2 | 116 | |||
X03662.1 | - | 2 | 80 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
