siRNA Details
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033952.2 | NP_001029124.1 | 2 | 144 | ||
NM_001033953.2 | NP_001029125.1 | 2 | 144 | |||
NM_001741.2 | NP_001732.1 | 2 | 120 | |||
NR_125898.1 | 2 | 120 | ||||
XM_017018283.1 | XP_016873772.1 | 2 | 195 | |||
XM_017018284.1 | XP_016873773.1 | 2 | 219 | |||
GenBank mRNA | BC069684.1 | - | 2 | 120 | 7262564 | |
BC069704.1 | - | 2 | 120 | 7262481 | ||
BC069760.1 | - | 2 | 120 | 7262493 | ||
BC069778.1 | - | 2 | 120 | 7262505 | ||
BC093753.1 | - | 2 | 131 | 7939598 | ||
BC101599.1 | - | 2 | 107 | 8069105 | ||
BC143562.1 | - | 2 | 131 | 9052073 | ||
CD612754.1 | - | 1 | 53 | |||
M26095.1 | - | 2 | 103 | |||
M64486.1 | - | 2 | 108 | |||
X00356.1 | - | 2 | 139 | |||
X03662.1 | - | 2 | 103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
