siRNA Details
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006470.3 | NP_006461.3 | 9 | 2591 | ||
GenBank mRNA | AF096870.1 | - | 6 | 2259 | ||
AK002035.1 | - | 6 | 2057 | |||
AK056026.1 | - | 7 | 2873 | |||
BC001564.1 | - | 6 | 1921 | |||
BC015674.2 | - | 5 | 1609 | |||
BC053514.1 | - | 9 | 2591 | 6263729 | ||
BC067096.1 | - | 9 | 2591 | 6088764 |
Gene Symbol: ATP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000052.6 | NP_000043.4 | 23 | 5854 | ||
NM_001282224.1 | NP_001269153.1 | 22 | 5620 | |||
NR_104109.1 | 10 | 2863 | ||||
GenBank mRNA | L06133.1 | - | 23 | 5799 |
Gene Symbol: CLYBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011521051.2 | XP_011519353.1 | 9 | 2363 | ||
XM_017020402.1 | XP_016875891.1 | 8 | 2261 | |||
XM_017020404.1 | XP_016875893.1 | 8 | 3231 |
Gene Symbol: FAM213A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI589570.1 | - | 1 | 484 |
Gene Symbol: FNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251815.2 | XP_005251872.1 | 17 | 4453 | ||
XM_005251820.2 | XP_005251877.1 | 16 | 4399 | |||
XM_005251821.2 | XP_005251878.1 | 16 | 4396 | |||
XM_005251822.2 | XP_005251879.1 | 15 | 4384 | |||
XM_005251823.2 | XP_005251880.1 | 15 | 4381 | |||
XM_005251825.2 | XP_005251882.1 | 16 | 4366 | |||
XM_005251827.2 | XP_005251884.1 | 15 | 4354 | |||
XM_005251828.2 | XP_005251885.1 | 15 | 4351 | |||
XM_005251829.2 | XP_005251886.1 | 16 | 4309 | |||
XM_005251830.2 | XP_005251887.1 | 15 | 4297 | |||
XM_005251831.2 | XP_005251888.1 | 15 | 4216 | |||
XM_005251832.2 | XP_005251889.1 | 14 | 4201 | |||
XM_005251833.2 | XP_005251890.1 | 14 | 4198 | |||
XM_006717016.2 | XP_006717079.1 | 17 | 4396 | |||
XM_011518399.2 | XP_011516701.1 | 17 | 4483 | |||
XM_011518400.2 | XP_011516702.1 | 17 | 4480 | |||
XM_017014487.1 | XP_016869976.1 | 17 | 4393 | |||
XM_017014488.1 | XP_016869977.1 | 16 | 4369 | |||
XM_017014489.1 | XP_016869978.1 | 16 | 4312 | |||
XM_017014492.1 | XP_016869981.1 | 17 | 4769 | |||
XM_017014493.1 | XP_016869982.1 | 15 | 4213 |
Gene Symbol: FXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000144.4 | NP_000135.2 | 5 | 5155 | ||
NM_181425.2 | NP_852090.1 | 5 | 5163 |
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243650.1 | NP_001230579.1 | 2 | 1448 |
Gene Symbol: HACD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005855.1 | XP_016861344.1 | 6 | 2261 | ||
GenBank mRNA | AL832532.1 | - | 3 | 2050 | ||
AY191814.1 | - | 7 | 2592 | |||
BC060839.1 | - | 7 | 2481 | 5273705 |
Gene Symbol: LOC100506486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295063.1 | - | 1 | 479 |
Gene Symbol: LOC101929199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125921.1 | 6 | 976 | |||
GenBank mRNA | AK125145.1 | - | 6 | 976 |
Gene Symbol: LOC105372321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627135.1 | - | 1 | 3058 |
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK297498.1 | - | 4 | 783 | ||
AK310573.1 | - | 4 | 560 | |||
BX647356.1 | - | 2871 |
Gene Symbol: RPGRIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255867.1 | XP_005255924.1 | 22 | 3350 | ||
XM_017023094.1 | XP_016878583.1 | 24 | 3464 | |||
XM_017023098.1 | XP_016878587.1 | 10 | 2427 | |||
XM_017023099.1 | XP_016878588.1 | 10 | 1793 | |||
GenBank mRNA | AK307929.1 | - | 22 | 3370 |
Gene Symbol: SIGLEC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY854038.1 | - | 7 | 1699 |
Gene Symbol: TRIM16L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037330.1 | NP_001032407.1 | 5 | 1666 | ||
XM_005256479.4 | XP_005256536.1 | 8 | 1840 | |||
XM_005256480.4 | XP_005256537.1 | 9 | 1944 | |||
XM_005256481.4 | XP_005256538.1 | 11 | 2193 | |||
XM_005256483.4 | XP_005256540.1 | 10 | 2085 | |||
XM_011523689.2 | XP_011521991.1 | 6 | 1545 | |||
XM_011523690.2 | XP_011521992.1 | 12 | 2255 | |||
XM_017024245.1 | XP_016879734.1 | 13 | 2555 | |||
XM_017024246.1 | XP_016879735.1 | 12 | 2417 | |||
XM_017024247.1 | XP_016879736.1 | 13 | 2558 | |||
XM_017024248.1 | XP_016879737.1 | 12 | 2447 | |||
XM_017024249.1 | XP_016879738.1 | 11 | 2309 | |||
XM_017024250.1 | XP_016879739.1 | 11 | 2310 | |||
XM_017024251.1 | XP_016879740.1 | 11 | 2147 | |||
XM_017024252.1 | XP_016879741.1 | 6 | 1801 | |||
GenBank mRNA | AK298836.1 | - | 5 | 1611 | ||
AL080124.1 | - | 6 | 560 | |||
DQ232882.1 | - | 5 | 1666 |
Gene Symbol: ZFP69B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271137.2 | XP_005271194.1 | 6 | 2381 | ||
XM_005271139.3 | XP_005271196.1 | 5 | 2329 |
Gene Symbol: ZNF844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026664.1 | XP_016882153.1 | 3 | 2880 | ||
GenBank mRNA | AK027865.1 | - | 1 | 2319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
