siRNA Details
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022814.1 | XP_016878303.1 | 8 | 1396 | ||
XM_017022815.1 | XP_016878304.1 | 8 | 916 | |||
XM_017022816.1 | XP_016878305.1 | 8 | 1396 | |||
XM_017022817.1 | XP_016878306.1 | 7 | 1308 | |||
XM_017022818.1 | XP_016878307.1 | 7 | 2633 | |||
XM_017022819.1 | XP_016878308.1 | 8 | 1323 | |||
XM_017022820.1 | XP_016878309.1 | 8 | 1040 | |||
XM_017022821.1 | XP_016878310.1 | 7 | 1906 | |||
XM_017022822.1 | XP_016878311.1 | 8 | 2909 | |||
XM_017022823.1 | XP_016878312.1 | 6 | 2545 | |||
XM_017022826.1 | XP_016878315.1 | 7 | 857 | |||
XM_017022827.1 | XP_016878316.1 | 3 | 604 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023622.1 | XP_016879111.1 | 7 | 832 | ||
XM_017023623.1 | XP_016879112.1 | 7 | 832 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144350.1 | - | 6 | 603 | 9052870 | |
BX647358.1 | - | 2 | 3978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
