siRNA Details
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 5, 6 | 630 | ||
GenBank mRNA | AB209632.1 | - | 4, 5 | 3197 | ||
AK294452.1 | - | 1, 2 | 193 | |||
AK295170.1 | - | 5, 6 | 629 | |||
AK303500.1 | - | 1, 2 | 124 | |||
AK304162.1 | - | 3, 4 | 412 | |||
AK316459.1 | - | 1, 2 | 59 | |||
BC036263.1 | - | 5, 6 | 630 | 3903371 | ||
BC094882.1 | - | 4, 5 | 557 | 3093162 | ||
BM557638.1 | - | 1, 2 | 204 | |||
KF175175.1 | - | 4, 5 | 497 | |||
KF175176.1 | - | 4, 5 | 554 | |||
KF175177.1 | - | 4, 5 | 554 | |||
KF175178.1 | - | 4, 5 | 455 | |||
KF175179.1 | - | 4, 5 | 497 | |||
KF175180.1 | - | 4, 5 | 497 | |||
KF175181.1 | - | 4, 5 | 641 | |||
KF175182.1 | - | 4, 5 | 497 | |||
KF175183.1 | - | 3, 4 | 389 | |||
KF175184.1 | - | 3, 4 | 380 | |||
KF175185.1 | - | 4, 5 | 497 | |||
KF175186.1 | - | 3, 4 | 371 | |||
KF175187.1 | - | 4, 5 | 497 | |||
KF175188.1 | - | 4, 5 | 458 | |||
KF175189.1 | - | 4, 5 | 458 | |||
KF175190.1 | - | 4, 5 | 497 | |||
KF175191.1 | - | 4, 5 | 458 | |||
KF175192.1 | - | 4, 5 | 458 | |||
KF175193.1 | - | 4, 5 | 461 | |||
KF175194.1 | - | 4, 5 | 461 | |||
KF175196.1 | - | 4, 5 | 458 | |||
KF175197.1 | - | 4, 5 | 458 | |||
KF175198.1 | - | 4, 5 | 554 | |||
KF175199.1 | - | 4, 5 | 554 | |||
KF175200.1 | - | 4, 5 | 554 | |||
KF175201.1 | - | 4, 5 | 497 | |||
KF175202.1 | - | 4, 5 | 458 | |||
KF175203.1 | - | 4, 5 | 554 | |||
KF175205.1 | - | 4, 5 | 554 | |||
KF175206.1 | - | 4, 5 | 554 | |||
KF175215.1 | - | 6, 7 | 816 | |||
KF175216.1 | - | 5, 6 | 831 | |||
KF175218.1 | - | 4, 5 | 459 | |||
KF175223.1 | - | 4, 5 | 554 | |||
KF175224.1 | - | 4, 5 | 497 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 6, 7 | 747 | ||
XM_017023514.1 | XP_016879003.1 | 6, 7 | 1036 | |||
XM_017023515.1 | XP_016879004.1 | 7, 8 | 1055 | |||
XM_017023516.1 | XP_016879005.1 | 6, 7 | 1082 | |||
XM_017023517.1 | XP_016879006.1 | 6, 7 | 813 | |||
XM_017023518.1 | XP_016879007.1 | 6, 7 | 901 | |||
XM_017023519.1 | XP_016879008.1 | 6, 7 | 958 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 4, 5 | 554 | ||
GenBank mRNA | KF175195.1 | - | 4, 5 | 458 | ||
KF175204.1 | - | 4, 5 | 497 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 4, 5 | 576 | ||
XM_011523496.2 | XP_011521798.1 | 5, 6 | 632 | |||
XM_011523498.2 | XP_011521800.1 | 5, 6 | 666 | |||
XM_017023248.1 | XP_016878737.1 | 5, 6 | 1069 | |||
XM_017023249.1 | XP_016878738.1 | 4, 5 | 886 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 5, 6 | 779 | ||
XM_017023244.1 | XP_016878733.1 | 4, 5 | 2790 | |||
XM_017023245.1 | XP_016878734.1 | 5, 6 | 2627 | |||
XM_017023246.1 | XP_016878735.1 | 5, 6 | 2548 | |||
XM_017023247.1 | XP_016878736.1 | 4, 5 | 781 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 4, 5 | 833 | ||
GenBank mRNA | AK296301.1 | - | 114 | |||
AK296338.1 | - | 4, 5 | 674 | |||
AK303264.1 | - | 1, 2 | 146 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 4, 5 | 646 | ||
XM_005255741.3 | XP_005255798.1 | 5, 6 | 1458 | |||
XM_005255742.4 | XP_005255799.1 | 5, 6 | 1204 | |||
XM_011545929.2 | XP_011544231.1 | 6, 7 | 1278 | |||
XM_011545931.2 | XP_011544233.1 | 5, 6 | 911 | |||
XM_017023628.1 | XP_016879117.1 | 5, 6 | 661 | |||
XM_017023629.1 | XP_016879118.1 | 5, 6 | 1204 | |||
XM_017023630.1 | XP_016879119.1 | 6, 7 | 931 | |||
XM_017023631.1 | XP_016879120.1 | 4, 5 | 1275 | |||
XM_017023632.1 | XP_016879121.1 | 5, 6 | 661 | |||
GenBank mRNA | BC144439.1 | - | 4, 5 | 607 | ||
BC157836.1 | - | 4, 5 | 1296 | 9057055 | ||
KF175222.1 | - | 4, 5 | 608 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 4, 5 | 608 | ||
XM_017023622.1 | XP_016879111.1 | 5, 6 | 732 | |||
XM_017023623.1 | XP_016879112.1 | 5, 6 | 732 | |||
XM_017023624.1 | XP_016879113.1 | 5, 6 | 732 | |||
XM_017023625.1 | XP_016879114.1 | 5, 6 | 712 | |||
XM_017023626.1 | XP_016879115.1 | 6, 7 | 1351 | |||
XM_017023627.1 | XP_016879116.1 | 5, 6 | 732 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 4, 5 | 646 | ||
NM_001287251.1 | NP_001274180.1 | 5, 6 | 732 | |||
XM_003118719.7 | XP_003118767.2 | 5, 6 | 1207 | |||
XM_011545690.2 | XP_011543992.1 | 6, 7 | 1296 | |||
XM_011545693.2 | XP_011543995.1 | 6, 7 | 1295 | |||
XM_017022814.1 | XP_016878303.1 | 6, 7 | 1296 | |||
XM_017022815.1 | XP_016878304.1 | 6, 7 | 816 | |||
XM_017022816.1 | XP_016878305.1 | 6, 7 | 1296 | |||
XM_017022817.1 | XP_016878306.1 | 5, 6 | 1208 | |||
XM_017022818.1 | XP_016878307.1 | 5, 6 | 2533 | |||
XM_017022819.1 | XP_016878308.1 | 6, 7 | 1223 | |||
XM_017022820.1 | XP_016878309.1 | 6, 7 | 940 | |||
XM_017022821.1 | XP_016878310.1 | 5, 6 | 1806 | |||
XM_017022822.1 | XP_016878311.1 | 6, 7 | 2809 | |||
XM_017022823.1 | XP_016878312.1 | 4, 5 | 2445 | |||
XM_017022824.1 | XP_016878313.1 | 6, 7 | 1222 | |||
XM_017022825.1 | XP_016878314.1 | 6, 7 | 940 | |||
XM_017022826.1 | XP_016878315.1 | 5, 6 | 757 | |||
XM_017022828.1 | XP_016878317.1 | 6, 7 | 1295 | |||
GenBank mRNA | BC171825.1 | - | 5, 6 | 1382 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063633.1 | - | 21, 22 | 2147 | ||
BC144350.1 | - | 4, 5 | 503 | 9052870 | ||
KF175207.1 | - | 4, 5 | 503 | |||
KF175220.1 | - | 4, 5 | 408 | |||
KF175221.1 | - | 4, 5 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
