siRNA Details
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545690.2 | XP_011543992.1 | 9 | 1428 | ||
XM_017022814.1 | XP_016878303.1 | 8 | 1503 | |||
XM_017022815.1 | XP_016878304.1 | 8 | 1023 | |||
XM_017022816.1 | XP_016878305.1 | 8 | 1503 | |||
XM_017022817.1 | XP_016878306.1 | 7 | 1415 | |||
XM_017022818.1 | XP_016878307.1 | 7 | 2740 | |||
XM_017022819.1 | XP_016878308.1 | 8 | 1430 | |||
XM_017022820.1 | XP_016878309.1 | 8 | 1147 | |||
XM_017022821.1 | XP_016878310.1 | 7 | 2013 | |||
XM_017022822.1 | XP_016878311.1 | 8 | 3016 | |||
XM_017022823.1 | XP_016878312.1 | 6 | 2652 | |||
XM_017022826.1 | XP_016878315.1 | 7 | 964 | |||
XM_017022827.1 | XP_016878316.1 | 3 | 711 | |||
XM_017022828.1 | XP_016878317.1 | 9 | 1427 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255742.4 | XP_005255799.1 | 8 | 1336 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023622.1 | XP_016879111.1 | 7 | 939 | ||
XM_017023623.1 | XP_016879112.1 | 7 | 939 | |||
XM_017023627.1 | XP_016879116.1 | 8 | 864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
