siRNA Details
Gene Symbol: CSGALNACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023822.1 | - | 1 | 3814 |
Gene Symbol: DROSHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209501.1 | - | 16 | 11559 |
Gene Symbol: KIAA1217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024199.1 | - | 1 | 221 |
Gene Symbol: NMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004808.2 | NP_004799.1 | 12 | 3142 | ||
XM_006717539.3 | XP_006717602.1 | 11 | 2979 | |||
XM_017016951.1 | XP_016872440.1 | 12 | 2946 | |||
GenBank mRNA | AK123732.1 | - | 1 | 208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
