siRNA Details
Gene Symbol: ZNF266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095928.1 | - | 1 | 1058 |
Gene Symbol: EIF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209364.1 | - | 1 | 2145 |
Gene Symbol: EVI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308248.1 | NP_001295177.1 | 19 | 3198 | ||
NM_005665.5 | NP_005656.4 | 18 | 3165 | |||
XM_017002272.1 | XP_016857761.1 | 21 | 3424 | |||
GenBank mRNA | AB209513.1 | - | 15 | 2351 | ||
AF008915.1 | - | 19 | 3131 |
Gene Symbol: FOXL2NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040061.2 | NP_001035150.1 | 3 | 3331 | ||
XM_005247443.3 | XP_005247500.1 | 4 | 3417 | |||
GenBank mRNA | AK125319.1 | - | 3 | 3331 |
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 24 | 4047 | ||
XM_005259896.3 | XP_005259953.1 | 24 | 4086 | |||
GenBank mRNA | BM561964.1 | - | 1 | 755 | ||
U09607.1 | - | 23 | 4045 |
Gene Symbol: MAMSTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026642.1 | XP_016882131.1 | 7 | 2230 |
Gene Symbol: PLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091897.1 | - | 6 | 927 |
Gene Symbol: SARNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222719.1 | - | 7 | 1729 |
Gene Symbol: SEMA4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126512.1 | - | 1 | 820 |
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648303.1 | - | 2 | 2149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
