siRNA Details
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 30 | 3859 | ||
NM_001291398.1 | NP_001278327.1 | 29 | 3796 | |||
XM_005269806.3 | XP_005269863.2 | 29 | 3735 | |||
XM_005269807.3 | XP_005269864.2 | 29 | 3720 | |||
XM_005269808.3 | XP_005269865.2 | 28 | 3621 | |||
XM_005269810.3 | XP_005269867.2 | 12 | 1952 | |||
XM_006717831.3 | XP_006717894.2 | 29 | 3654 | |||
XM_006717832.3 | XP_006717895.2 | 27 | 3519 | |||
XM_011539786.2 | XP_011538088.1 | 27 | 3408 | |||
XM_017016227.1 | XP_016871716.1 | 29 | 3684 | |||
XM_017016228.1 | XP_016871717.1 | 28 | 3672 | |||
XM_017016229.1 | XP_016871718.1 | 28 | 3657 | |||
XM_017016230.1 | XP_016871719.1 | 29 | 3672 | |||
XM_017016231.1 | XP_016871720.1 | 28 | 3591 | |||
XM_017016232.1 | XP_016871721.1 | 27 | 3585 | |||
XM_017016233.1 | XP_016871722.1 | 27 | 3382 | |||
XM_017016234.1 | XP_016871723.1 | 29 | 3742 | |||
XM_017016235.1 | XP_016871724.1 | 21 | 2758 | |||
GenBank mRNA | AK125147.1 | - | 2 | 2213 | ||
AK304116.1 | - | 30 | 3795 | |||
AL080183.1 | - | 6 | 1191 | |||
BC065500.1 | - | 2 | 97 | |||
BC075815.1 | - | 26 | 3361 | |||
BC082258.1 | - | 30 | 3786 | 6278433 | ||
BC144479.1 | - | 29 | 3825 | 9053006 | ||
BQ060891.1 | - | 2 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
