siRNA Details
Gene Symbol: SLC6A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142805.1 | NP_001136277.1 | 13 | 2185 | ||
NM_001142806.1 | NP_001136278.1 | 13 | 1764 | |||
NM_005629.3 | NP_005620.1 | 13 | 2215 | |||
GenBank mRNA | AB209704.1 | - | 13 | 1989 | ||
AK295495.1 | - | 13 | 1764 | |||
BC012355.1 | - | 13 | 1982 | 4634609 | ||
BC081558.1 | - | 13 | 2216 | 4823748 | ||
EU280316.1 | - | 9 | 1976 | |||
L31409.1 | - | 13 | 2410 | |||
S74039.1 | - | 13 | 1955 | |||
U17986.1 | - | 6 | 2257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
