siRNA Details
Gene Symbol: SLC6A10PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC068290.1 | - | 3 | 412 |
Gene Symbol: SLC6A10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003083.2 | 8 | 3445 | |||
GenBank mRNA | AK125843.1 | - | 8 | 3446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
