siRNA Details
Gene Symbol: GNRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310220.1 | NP_001297149.1 | 2 | 62 | ||
NM_001501.1 | NP_001492.1 | 2 | 67 | |||
NM_178331.1 | NP_847901.1 | 2 | 67 | |||
NM_178332.1 | NP_847902.1 | 2 | 67 | |||
XM_017027823.1 | XP_016883312.1 | 2 | 2674 | |||
GenBank mRNA | AF036330.1 | - | 2 | 67 | ||
BC069362.1 | - | 2 | 66 | 7262673 | ||
BC115399.1 | - | 2 | 66 | 40069240 | ||
BC115400.1 | - | 2 | 61 | 40069241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
