siRNA Details
Gene Symbol: ZFPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL389989.1 | - | 1 | 608 |
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126043.1 | - | 1 | 2175 |
Gene Symbol: CADM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256502.1 | NP_001243431.1 | 1 | 82 | ||
NM_001256503.1 | NP_001243432.1 | 1 | 82 | |||
NM_001256504.1 | NP_001243433.1 | 1 | 82 | |||
NM_001256505.1 | NP_001243434.1 | 1 | 82 | |||
GenBank mRNA | AB646740.1 | - | 1 | 82 | ||
AB646741.1 | - | 1 | 82 | |||
AB646742.1 | - | 1 | 82 | |||
AB646743.1 | - | 1 | 82 |
Gene Symbol: CCDC129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047565.1 | 1 | 412 | |||
GenBank mRNA | AK128026.1 | - | 1 | 412 |
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB095932.1 | - | 1 | 1082 |
Gene Symbol: LOC102724058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110598.1 | 1 | 115 | |||
GenBank mRNA | HY008183.1 | - | 1 | 92 | ||
HY011842.1 | - | 1 | 115 | |||
HY029566.1 | - | 1 | 103 |
Gene Symbol: LOC107986533 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011597.1 | XP_016867086.1 | 2 | 463 |
Gene Symbol: LOC283585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038445.1 | 1 | 118 | |||
GenBank mRNA | BC034577.1 | - | 1 | 118 |
Gene Symbol: MDGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC142947.1 | - | 1 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
