siRNA Details
Gene Symbol: KRTAP3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031958.1 | NP_114164.1 | 1 | 185 | ||
GenBank mRNA | AJ406931.1 | - | 1 | 185 | ||
BC113077.1 | - | 1 | 165 | 40069149 | ||
BC113078.1 | - | 1 | 169 | 40069150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
