siRNA Details
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 282 |
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3901 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 2967 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3768 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3455 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 4030 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4294 | |||
NM_003879.5 | NP_003870.4 | 10 | 3873 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4768 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3737 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3816 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4703 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6720 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4576 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 3914 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3539 |
Gene Symbol: ERVK-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ304053.1 | - | 1 | 65 |
Gene Symbol: FKSG48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336875.1 | - | 1 | 276 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 201 | ||
AF336879.1 | - | 1 | 225 | |||
AF336882.1 | - | 1 | 261 | |||
AF336883.1 | - | 1 | 210 | |||
AF336884.1 | - | 1 | 270 | |||
AF336886.1 | - | 1 | 198 | |||
AF338190.1 | - | 1 | 243 | |||
AF338193.1 | - | 1 | 261 | |||
AF338194.1 | - | 1 | 264 | |||
AF338195.1 | - | 1 | 261 | |||
AF338196.1 | - | 1 | 273 | |||
AF338198.1 | - | 1 | 261 | |||
AF469044.1 | - | 1 | 210 |
Gene Symbol: HELLPAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JX088243.1 | - | 1 | 53409 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 273 | ||
AF336881.1 | - | 1 | 273 | |||
AF338191.1 | - | 1 | 258 |
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 2323 | ||
NM_145280.5 | NP_660323.3 | 4 | 2184 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 22 | 2907 | ||
GenBank mRNA | BC063885.1 | - | 22 | 2893 | 30383658 |
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209437.1 | - | 1 | 414 |
Gene Symbol: SPICE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832327.2 | - | 12 | 3048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
