siRNA Details
Gene Symbol: MAGEA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011543.2 | NP_001011543.2 | 5 | 2263 | ||
NM_001251828.1 | NP_001238757.1 | 5 | 2191 | |||
NM_021048.4 | NP_066386.2 | 4 | 2172 |
Gene Symbol: LOC100147773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124964.1 | - | 2 | 1563 |
Gene Symbol: PTGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311280.1 | - | 4 | 452 |
Gene Symbol: SYNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JQ740785.1 | - | 3 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
