siRNA Details
Gene Symbol: LILRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081450.2 | NP_001074919.2 | 13 | 2246 | ||
NM_001320960.1 | NP_001307889.1 | 14 | 2294 | |||
NM_006864.3 | NP_006855.3 | 13 | 2243 | |||
NR_135493.1 | 13 | 2271 | ||||
NR_135494.1 | 12 | 2104 | ||||
NR_135495.1 | 13 | 2163 | ||||
NR_135496.1 | 14 | 2326 | ||||
GenBank mRNA | BC028153.1 | - | 14 | 2217 |
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004337.1 | XP_016859826.1 | 1 | 2328 | ||
XM_017004338.1 | XP_016859827.1 | 1 | 2328 |
Gene Symbol: CACNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006078.3 | NP_006069.1 | 4 | 1174 | ||
XM_017028531.1 | XP_016884020.1 | 3 | 971 |
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 5 | 2506 | ||
NM_020351.3 | NP_065084.2 | 4 | 2444 | |||
GenBank mRNA | AY550974.1 | - | 3 | 2307 | ||
BC013581.1 | - | 5 | 2484 | 3875911 | ||
BG938623.1 | - | 1 | 318 | |||
CB269463.1 | - | 1 | 32 |
Gene Symbol: DACOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY203937.1 | - | 2 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
