siRNA Details
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005645.3 | NP_005636.1 | 3 | 198 | ||
GenBank mRNA | AK312158.1 | - | 3 | 194 | ||
AU280379.1 | - | 3 | 198 | |||
AV713996.1 | - | 3 | 145 | |||
BC121180.1 | - | 3 | 172 | 40123243 | ||
BC121181.1 | - | 3 | 172 | 40123247 | ||
X84003.1 | - | 3 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
