siRNA Details
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 5, 6 | 634 | ||
NM_001286835.1 | NP_001273764.1 | 5, 6 | 634 | |||
XM_005272748.1 | XP_005272805.1 | 5, 6 | 594 | |||
XM_005272749.1 | XP_005272806.1 | 5, 6 | 594 | |||
XM_005272751.1 | XP_005272808.1 | 5, 6 | 594 | |||
XM_006716914.1 | XP_006716977.1 | 5, 6 | 594 | |||
XM_017014455.1 | XP_016869944.1 | 5, 6 | 594 | |||
XM_017014456.1 | XP_016869945.1 | 6, 7 | 525 | |||
XM_017014457.1 | XP_016869946.1 | 6, 7 | 525 | |||
XM_017014458.1 | XP_016869947.1 | 5, 6 | 594 | |||
XM_017014459.1 | XP_016869948.1 | 5, 6 | 540 | |||
XM_017014460.1 | XP_016869949.1 | 5, 6 | 594 | |||
XM_017014461.1 | XP_016869950.1 | 5, 6 | 540 | |||
GenBank mRNA | AF065414.1 | - | 3, 4 | 251 | ||
AK097639.1 | - | 2, 3 | 682 | |||
AK292585.1 | - | 5, 6 | 574 | |||
BC067803.1 | - | 5, 6 | 572 | 5294979 | ||
BC082271.1 | - | 5, 6 | 530 | |||
BC141856.1 | - | 5, 6 | 572 | |||
DB463072.2 | - | 5, 6 | 572 |
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF257330.2 | - | 5, 6 | 555 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 5, 6 | 669 | ||
XM_005263600.2 | XP_005263657.1 | 5, 6 | 669 | |||
XM_005263602.2 | XP_005263659.1 | 5, 6 | 669 | |||
XM_005263603.2 | XP_005263660.1 | 6, 7 | 703 | |||
XM_005263604.2 | XP_005263661.1 | 5, 6 | 669 | |||
XM_005263605.3 | XP_005263662.1 | 6, 7 | 704 | |||
XM_011510667.1 | XP_011508969.1 | 5, 6 | 669 | |||
XM_011510668.2 | XP_011508970.1 | 2, 3 | 229 | |||
XM_011510669.2 | XP_011508971.1 | 5, 6 | 568 | |||
XM_017003407.1 | XP_016858896.1 | 5, 6 | 669 | |||
XM_017003408.1 | XP_016858897.1 | 6, 7 | 688 | |||
GenBank mRNA | AF452722.1 | - | 5, 6 | 525 | ||
AK225251.1 | - | 5, 6 | 538 | |||
AK290072.1 | - | 5, 6 | 669 | |||
AK303653.1 | - | 6, 7 | 708 | |||
AY343910.1 | - | 5, 6 | 554 | |||
BC120991.1 | - | 5, 6 | 540 | 40116409 | ||
BC120992.1 | - | 5, 6 | 540 | 40116411 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201453.3 | NP_958861.2 | 5, 6 | 578 | ||
XM_005277637.2 | XP_005277694.1 | 5, 6 | 657 | |||
XM_005277639.2 | XP_005277696.1 | 5, 6 | 657 | |||
XM_005277640.2 | XP_005277697.1 | 5, 6 | 657 | |||
XM_006717116.1 | XP_006717179.1 | 5, 6 | 657 | |||
XM_017014731.1 | XP_016870220.1 | 5, 6 | 3029 | |||
GenBank mRNA | AY344633.1 | - | 5, 6 | 554 | ||
BX509475.1 | - | 5, 6 | 578 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085457.1 | NP_001078926.1 | 5, 6 | 597 | ||
XM_017015029.1 | XP_016870518.1 | 1, 2 | 416 | |||
XM_017015035.1 | XP_016870524.1 | 5, 6 | 657 | |||
XM_017015036.1 | XP_016870525.1 | 5, 6 | 657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
