siRNA Details
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 7 | 722 | ||
NM_001145356.1 | NP_001138828.1 | 6 | 624 | |||
NM_018491.3 | NP_060961.3 | 6 | 624 | |||
XM_005251513.4 | XP_005251570.1 | 5 | 572 | |||
XM_005251514.1 | XP_005251571.1 | 6 | 624 | |||
XM_005251515.1 | XP_005251572.1 | 6 | 624 | |||
XM_005251517.4 | XP_005251574.1 | 6 | 624 | |||
XM_011517961.2 | XP_011516263.1 | 7 | 716 | |||
XM_011517962.2 | XP_011516264.1 | 6 | 624 | |||
XM_011517963.1 | XP_011516265.1 | 3 | 162 | |||
XM_011517964.2 | XP_011516266.1 | 6 | 624 | |||
XM_017014925.1 | XP_016870414.1 | 6 | 624 | |||
GenBank mRNA | AF212253.1 | - | 6 | 592 | ||
AF257330.2 | - | 6 | 582 | |||
AK290648.1 | - | 7 | 694 | |||
AK297292.1 | - | 7 | 663 | |||
AK315855.1 | - | 6 | 548 | |||
AY343911.1 | - | 6 | 581 | |||
BC005996.1 | - | 6 | 624 | 4069589 | ||
BC009573.1 | - | 7 | 675 | 4133536 | ||
BC013432.1 | - | 6 | 595 | 4133722 | ||
BC132759.1 | - | 6 | 558 | 40146781 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 6 | 696 | ||
XM_005263600.2 | XP_005263657.1 | 6 | 696 | |||
XM_005263602.2 | XP_005263659.1 | 6 | 696 | |||
XM_005263603.2 | XP_005263660.1 | 7 | 730 | |||
XM_005263604.2 | XP_005263661.1 | 6 | 696 | |||
XM_005263605.3 | XP_005263662.1 | 7 | 731 | |||
XM_011510667.1 | XP_011508969.1 | 6 | 696 | |||
XM_011510668.2 | XP_011508970.1 | 3 | 256 | |||
XM_011510669.2 | XP_011508971.1 | 6 | 595 | |||
XM_017003407.1 | XP_016858896.1 | 6 | 696 | |||
XM_017003408.1 | XP_016858897.1 | 7 | 715 | |||
GenBank mRNA | AF452722.1 | - | 6 | 552 | ||
AK225251.1 | - | 6 | 565 | |||
AK290072.1 | - | 6 | 696 | |||
AK303653.1 | - | 7 | 735 | |||
AY343910.1 | - | 6 | 581 | |||
BC120991.1 | - | 6 | 567 | 40116409 | ||
BC120992.1 | - | 6 | 567 | 40116411 |
Gene Symbol: CBWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291821.1 | NP_001278750.1 | 5 | 545 | ||
NM_201453.3 | NP_958861.2 | 6 | 605 | |||
XM_005277637.2 | XP_005277694.1 | 6 | 684 | |||
XM_005277639.2 | XP_005277696.1 | 6 | 684 | |||
XM_005277640.2 | XP_005277697.1 | 6 | 684 | |||
XM_006717116.1 | XP_006717179.1 | 6 | 684 | |||
XM_017014730.1 | XP_016870219.1 | 5 | 624 | |||
XM_017014731.1 | XP_016870220.1 | 6 | 3056 | |||
GenBank mRNA | AK297912.1 | - | 5 | 507 | ||
AY344633.1 | - | 6 | 581 | |||
BX509475.1 | - | 6 | 605 |
Gene Symbol: CBWD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024916.3 | NP_001020087.2 | 6 | 661 | ||
NM_001286835.1 | NP_001273764.1 | 6 | 661 | |||
XM_005272748.1 | XP_005272805.1 | 6 | 621 | |||
XM_005272749.1 | XP_005272806.1 | 6 | 621 | |||
XM_005272750.1 | XP_005272807.1 | 5 | 561 | |||
XM_005272751.1 | XP_005272808.1 | 6 | 621 | |||
XM_006716914.1 | XP_006716977.1 | 6 | 621 | |||
XM_017014455.1 | XP_016869944.1 | 6 | 621 | |||
XM_017014456.1 | XP_016869945.1 | 7 | 552 | |||
XM_017014457.1 | XP_016869946.1 | 7 | 552 | |||
XM_017014458.1 | XP_016869947.1 | 6 | 621 | |||
XM_017014459.1 | XP_016869948.1 | 6 | 567 | |||
XM_017014460.1 | XP_016869949.1 | 6 | 621 | |||
XM_017014461.1 | XP_016869950.1 | 6 | 567 | |||
GenBank mRNA | AF065414.1 | - | 4 | 278 | ||
AK097639.1 | - | 3 | 709 | |||
AK292585.1 | - | 6 | 601 | |||
BC067803.1 | - | 6 | 599 | 5294979 | ||
BC082271.1 | - | 6 | 557 | |||
BC141856.1 | - | 6 | 599 |
Gene Symbol: CBWD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085457.1 | NP_001078926.1 | 6 | 624 | ||
XM_017015019.1 | XP_016870508.1 | 5 | 624 | |||
XM_017015020.1 | XP_016870509.1 | 5 | 624 | |||
XM_017015021.1 | XP_016870510.1 | 5 | 624 | |||
XM_017015022.1 | XP_016870511.1 | 5 | 624 | |||
XM_017015023.1 | XP_016870512.1 | 5 | 624 | |||
XM_017015024.1 | XP_016870513.1 | 5 | 624 | |||
XM_017015025.1 | XP_016870514.1 | 5 | 624 | |||
XM_017015026.1 | XP_016870515.1 | 5 | 624 | |||
XM_017015027.1 | XP_016870516.1 | 5 | 624 | |||
XM_017015028.1 | XP_016870517.1 | 6 | 492 | |||
XM_017015029.1 | XP_016870518.1 | 2 | 443 | |||
XM_017015030.1 | XP_016870519.1 | 5 | 624 | |||
XM_017015032.1 | XP_016870521.1 | 5 | 624 | |||
XM_017015035.1 | XP_016870524.1 | 6 | 684 | |||
XM_017015036.1 | XP_016870525.1 | 6 | 684 | |||
GenBank mRNA | AK293173.1 | - | 5 | 531 | ||
BC069006.1 | - | 5 | 501 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146097, BC053745, AK076027, BC018472, AK080654, and others.
Gene Ontology Categories:
