siRNA Details
Gene Symbol: ELP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288726.1 | NP_001275655.1 | 9, 10 | 1207 | ||
GenBank mRNA | BC012514.1 | - | 9, 10 | 1158 | 4509903 |
Gene Symbol: C16orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022138.1 | - | 5 | 1727 |
Gene Symbol: CSAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126934.1 | - | 1 | 2622 |
Gene Symbol: HIST4H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020884.1 | - | 2 | 485 |
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012405.3 | NP_036537.1 | 5 | 3620 | ||
XM_011541140.2 | XP_011539442.1 | 4 | 3320 |
Gene Symbol: MIR194-2HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133638.1 | 2 | 936 |
Gene Symbol: PASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL117400.1 | - | 1 | 822 |
Gene Symbol: PDCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017004890.1 | XP_016860379.1 | 6 | 1599 |
Gene Symbol: TMEM116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020192.1 | XP_016875681.1 | 3 | 1598 |
Gene Symbol: TMEM169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142310.1 | NP_001135782.1 | 4 | 2209 | ||
NM_001142311.1 | NP_001135783.1 | 3 | 2123 | |||
NM_001142312.1 | NP_001135784.1 | 3 | 2117 | |||
NM_138390.3 | NP_612399.1 | 3 | 2119 | |||
XM_006712851.1 | XP_006712914.1 | 3 | 2323 | |||
XM_011512173.2 | XP_011510475.1 | 3 | 2514 | |||
GenBank mRNA | AK091582.1 | - | 3 | 2066 | ||
AK172731.1 | - | 1 | 1279 | |||
CR933667.1 | - | 3 | 2168 |
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017019922.1 | XP_016875411.1 | 3 | 1204 | ||
XM_017019923.1 | XP_016875412.1 | 3 | 1135 | |||
GenBank mRNA | AK122941.1 | - | 1 | 2245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
