siRNA Details
Gene Symbol: TCP11X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030010.1 | XP_016885499.1 | 6 | 685 | ||
XM_017030011.1 | XP_016885500.1 | 6 | 653 | |||
XM_017030012.1 | XP_016885501.1 | 5 | 632 | |||
XM_017030013.1 | XP_016885502.1 | 5 | 609 | |||
XM_017030014.1 | XP_016885503.1 | 6 | 654 |
Gene Symbol: NXF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK303069.1 | - | 6 | 691 |
Gene Symbol: TCP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093728.2 | NP_001087197.1 | 6 | 836 | ||
NM_001261817.1 | NP_001248746.1 | 6 | 821 | |||
NM_001261818.1 | NP_001248747.1 | 5 | 698 | |||
NM_001261819.1 | NP_001248748.1 | 5 | 683 | |||
NM_001261820.1 | NP_001248749.1 | 7 | 1042 | |||
NM_001261821.1 | NP_001248750.1 | 7 | 1027 | |||
NM_018679.5 | NP_061149.1 | 6 | 930 | |||
XM_005249339.2 | XP_005249396.1 | 5 | 591 | |||
XM_011514830.1 | XP_011513132.1 | 6 | 1068 | |||
XM_011514831.1 | XP_011513133.1 | 4 | 504 | |||
XM_011514832.1 | XP_011513134.1 | 3 | 368 | |||
GenBank mRNA | AF260330.1 | - | 3 | 379 | ||
AF269223.1 | - | 5 | 614 | |||
AF536532.1 | - | 7 | 882 | |||
AF536533.1 | - | 6 | 869 | |||
AK057234.1 | - | 6 | 653 | |||
AK098590.1 | - | 4 | 425 | |||
AK301778.1 | - | 6 | 712 | |||
AK301975.1 | - | 5 | 619 | |||
AK302022.1 | - | 5 | 584 | |||
AK302375.1 | - | 6 | 761 | |||
AK302456.1 | - | 6 | 742 | |||
AK315078.1 | - | 6 | 863 | |||
AK316403.1 | - | 6 | 645 | |||
AY069943.1 | - | 5 | 638 | |||
BC033729.1 | - | 6 | 782 | 5167179 | ||
BC040003.1 | - | 7 | 884 | 5741923 | ||
BC048418.1 | - | 6 | 860 | 5298037 | ||
BC090050.1 | - | 7 | 888 | 30387768 |
Gene Symbol: TCP11X2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277423.1 | NP_001264352.1 | 6 | 654 | ||
GenBank mRNA | AK093310.1 | - | 6 | 654 | ||
AK307893.1 | - | 6 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
