siRNA Details
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 1292 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 1099 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 1106 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 995 | |||
NM_013269.5 | NP_037401.1 | 5 | 1210 | |||
NR_036693.2 | 4 | 1181 | ||||
GenBank mRNA | AK310554.1 | - | 4 | 1172 | ||
BC094859.1 | - | 1 | 87 |
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539728.2 | XP_011538030.1 | 17 | 3276 | ||
XM_017016159.1 | XP_016871648.1 | 16 | 3208 |
Gene Symbol: FANCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029356.1 | XP_016884845.1 | 11 | 3475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
